Recommended storage conditions: temperatures from 5 to 30°C; humidity between 10 and 80%. The original sealed bag (if provided) must be kept intact before the first use to protect the purification element from foreign matter.
IQ70XXPKT1
Milli-Q® IQ Purification Kit
This product is replaced with IQ70XXPKIT. Search to Explore!
Synonym(s):
Milli-Q® IQ 7010-15 Purification kit
About This Item
Recommended Products
Quality Level
feature
Kit contains IPAK Gard® 10-15 Pretreatment Pack, IPAK Quanta® Polishing Cartridge and a Tank Vent Filter
packaging
pkg of 1 kit
compatibility
for use with Milli-Q® IQ 7010
for use with Milli-Q® IQ 7015
General description
- IPAK Gard® 10-15 Pretreatment Pack efficiently removes colloids, particles and free chlorine from tap water to protect the reverse osmosis (RO) membrane and improve system performance.
- IPAK Quanta® polishing cartridge contains innovative IQnano™ ion-exchange media which achieves ion removal down to trace levels. In addition, high-grade synthetic activated carbon targets trace organic contaminants.
- The Tank Vent Filter, redesigned for seamless integration, provides improved protection against airborne contaminants.
This kit is designed for use with Milli-Q® IQ 7010-15 systems for the production of pure (Type 2) and ultrapure (Type 1) water.
Application
Features and Benefits
- Activated carbon to protect against free chlorine.
- Depth filter to protect against particulates and colloids.
Tank Vent Filter contains:
- Activated carbon to remove volatile organics.
- Soda lime to remove CO2.
- Membrane filter to prevent particulates and bacteria from entering the tank.
IPAK Quanta® cartridge contains:
- Innovative IQnano™ ion-exchange media to remove ions down to trace levels.
- High-grade synthetic activated carbon targets trace organic contaminants.
In addition, 63% of the parts suppliers for the IPAK Gard® cartridge and 78% of the parts suppliers for IPAK Quanta® cartridge included in this kit are ISO 14001 environmental management system-certified.
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What are the storage conditions for this product?
1 answer-
Helpful?
-
-
How are consumables changed?
1 answer-
To help you to change your system's consumables, several video tutorials are available on our YouTube channel: https://youtube.com/playlist?list=PL2TUriaAaEA_IXDCnPbDk6OmrwptjwOHD. For high-flow systems, please keep in mind that all consumable changes must be approved by a 3-digit code that only the person in charge of the system knows.
Helpful?
-
-
How can I know which consumables can be used with my system?
1 answer-
To know which consumables can be used on your system, you can see in the user manual of your system or see all the consumables for each system listed on https://www.sigmaaldrich.com/products/water-purification/milli-q-consumables.
Helpful?
-
-
What steps are you taking towards improved sustainability?
1 answer-
Sustainability is at the heart of our innovations. From reducing water and energy consumption, to minimizing plastic and chemical waste, our dedicated water system engineers are constantly working on new ways to reduce the ecological footprint of our products to better support the sustainability goals we're all striving towards. For more information, we invite you to review the following brochure and infographic which highlight our efforts and achievements: https://www.sigmaaldrich.com/life-science/milliq#sus. Do not hesitate to contact your local representative or sales contact (https://www.sigmaaldrich.com/collections/offices) for further information.
Helpful?
-
-
Can I arrange to have my consumables automatically ordered and delivered once a year?
1 answer-
It's possible to place only one consumable order per year with a specific contract called "Consumable Supply Agreement (CSA)" (https://www.sigmaaldrich.com/US/en/products/water-purification/milli-q-consumables#csa). For further information, contact your local representative or sales contact (https://www.sigmaaldrich.com/US/en/collections/offices).
Helpful?
-
-
How frequently do the various consumables need to be changed?
1 answer-
The frequency that consumables must be changed depends on the system's usage (treated water volume) and on the consumables themselves (time of usage). In order to ensure the quality of water and your results, the consumables should be replaced when the system alerts you.
Helpful?
-
-
What is the consumables delivery time?
1 answer-
Refer to the consumable's Product page for a first estimation of its expected delivery time and contact customer service for more information.
Helpful?
-
-
If I have a final filter that I use only occasionally (e.g., Biopak®) and have another filter (e.g. Millipak®) that I most of the time, can I keep the 'occasional-use' filter for use again at a later time?
1 answer-
It is not possible to re-use a filter that has been used and then removed because air intake during replacement creates an airlock and blocks water passing through the filter. If you regularly need two different filters, we recommend using two POD dispensers.
Helpful?
-
-
Where can I find data sheets, Certificates of Origin, Certificates of Analysis, and Certificates of Quality?
1 answer-
Documents and certificates are available in the "Documentation" section on this page. Do not hesitate to contact us if you require further information.
Helpful?
-
-
Can consumables be opened and refilled?
1 answer-
Unfortunately, this is not possible because of safety issues and the need to guarantee the quality of consumables and their interior purification components. Our consumables are manufactured in an ISO 9001-certified facility France in agreement with high quality standards.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service