BI0222
Capping Reagent A THF/Pyridine/Acetic Anhydride
Biosynthesis Novabiochem®
Synonym(s):
dna
About This Item
Recommended Products
Quality Level
product line
Novabiochem®
form
liquid
reaction suitability
reaction type: Acetylations
reagent type: derivatization reagent
transition temp
flash point -21.5 °C (THF)
shipped in
ambient
storage temp.
2-30°C
Related Categories
Application
BI0222-0450 is packaged in 28/405 glass bottles
BI0221-4000 is packaged in 38/430 glass bottles
DNA Reagents Label Change Explanation
Analysis Note
Assay (pyridine): 9.5-10.5% v/v
Assay (tetrahydrofuran): 79-81% v/v
Color (APHA): 10 max
Form: Clear liquid
Water (H2O): 300ppm max
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
-6.7 °F - closed cup - (THF)
Flash Point(C)
-21.5 °C - closed cup - (THF)
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
PDSCL
Deleterious substance
PRTR
Class I Designated Chemical Substances
FSL
Group 4: Flammable liquids
Type 1 petroleums
Hazardous rank II
Water insoluble liquid
ISHL Indicated Name
Substances Subject to be Indicated Names
ISHL Notified Names
Substances Subject to be Notified Names
JAN Code
BI0222-0201:
BI0222-4000:
BI0222SE-37S:
BI0222-0450:
BI0222-NP20:
BI0222LT-38SK:
BI0222ES-37S:
BI0222SE-38S:
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service