430-30
Centriprep Centrifugal Filter Unit
30 kDa cutoff, 15 mL volume
About This Item
Recommended Products
material
Nylon cap
polycarbonate collector (for filtrate)
polycarbonate reservoir (for sample)
polypropylene seal
regenerated cellulose membrane
sterility
non-sterile
feature
holdup volume 0.5 mL
manufacturer/tradename
Centriprep®
parameter
15 mL sample volume
technique(s)
protein extraction: suitable
protein purification: suitable
L
135 cm
diam.
2.81 cm
filtration area
2.84 cm2
working volume
15 mL
pore size
30 kDa NMWCO
application(s)
sample preparation
shipped in
ambient
General description
Features & Benefits:
•For use with samples containing high levels of solutes
•Unique inverse flow mode of operation with large deadstop
•Fast sample processing
•Available with Ultracel membranes: (3, 10, 30 and 50 kDa NMWL)
•Fits standard swinging-bucket rotor for 50 mL tubes
Applications:
Concentration and Purification of Particle-laden Solutions, Solutions with High Solute Concentration, Separation of Low MW Solutes from Fermentation Broths, Cell Culture Media and Lysates
Application
Features and Benefits
Physical form
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service