Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

328006-M

Millipore

ERK Inhibitor

The ERK Inhibitor, also referenced under CAS 1049738-54-6, controls the biological activity of ERK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

ERK Inhibitor, 3-(2-Aminoethyl)-5-((4-ethoxyphenyl)methylene)-2,4-thiazolidinedione, HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16N2O3S · xHCl
CAS Number:
Molecular Weight:
292.35 (free base basis)
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

light yellow

solubility

DMSO: 10 mg/mL

storage temp.

2-8°C

SMILES string

S1C(=O)N(C(=O)C1=Cc2ccc(cc2)OCC)CCN.Cl

InChI key

PQVLWVGMXJPJLG-UHFFFAOYSA-N

General description

A cell-permeable and reversible thiazolidinedione compound that displays anti-proliferative properties (IC50 ≤ 25 µM in HeLa, A549, and SUM-159 tumor cells). Preferentially binds to ERK2 with a KD of ~5 µM and prevents its interaction with protein substrates. Shown to block ERK-mediated phosphorylation of ribosomal S6 kinase-1 (RSK-1) and ternary complex factor Elk-1 in HeLa cells, while exhibiting little effect on ERK1/2 phosphorylation by MEK1/2.

Biochem/physiol Actions

Primary Target
ERK
Target IC50: ≤ 25 µM for anti-proliferative properties in HeLa, A549, and SUM-159 tumor cells; KD of ~5 µM for ERK2

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Hancock, C. N., et al. 2005. J. Med. Chem.48, 4586.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

328006-1.1ML:
328006-MG:
328006-5MG-M:
328006-5MG:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service