Skip to Content
Merck
All Photos(1)

Documents

Safety Information

251765

Sigma-Aldrich

DAR-4M AM

A cell-permeable, photo-stable nitric oxide (NO) fluorescent indicator with a detection limit of ~10 nM.

Synonym(s):

DAR-4M AM, Diaminorhodamine-4M AM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H31IN4O5
Molecular Weight:
630.47
UNSPSC Code:
12352202
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

solution

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

dark red

fluorescence

λex ~560 nm
λem ~575 nm

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable, photo-stable nitric oxide (NO) fluorescent indicator with a detection limit of ~10 nM. Intracellularly releases DAR-4M (Cat. No. 251760) by the action of esterases. Reacts with NO, in the presence of oxygen, resulting in a triazolo-rhodamine analog (DAR-4M T) that exhibits about 840-fold greater fluorescence quantum efficiency. The fluorescence intensity is not dependent on pH above 4.0. Excitation max: ~560 nm; emission max: ~575 nm.
A cell-permeable, photo-stable nitric oxide (NO) fluorescent indicator with a detection limit of ~10 nM. Intracellularly releases DAR-4M (Cat. No. 251760) by the action of esterases. Reacts with NO, in the presence of O2, resulting in a triazolo-rhodamine analog (DAR-4M T) that exhibits about 840-fold greater fluorescence quantum efficiency. The fluorescence intensity is not dependent on pH.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Nitric oxide (NO) fluorescent indicator
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

In DMSO.

Reconstitution

Following initial thaw, aliquot and freeze at (-20°C). Use immediately after dilution, discard any unused portion.

Other Notes

Kojima, H., et al. 2001. Anal. Chem.73, 1967.
Kojima, H., et al. 2000. Tetrahedron Lett.41, 69.

Legal Information

Not available for sale in Japan.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water soluble liquid

JAN Code

251765-MG:
251765-1MG:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service