Skip to Content
Merck
All Photos(1)

Key Documents

14-743-M

Sigma-Aldrich

GCK Protein, active, 10 µg

Active, N-terminal GST-tagged, recombinant, human GCK amino acids 1-473, expressed by baculovirus in Sf21 cells. For use in Kinase Assays.

Synonym(s):

germinal center kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

specific activity

2337 U/mg

mol wt

Mw 80.6 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

General description

N-terminal GST- tagged, recombinant, human GCK amino acids 1-473, expressed by baculovirus in Sf21 cells.
Product Source: Recombinant protein expressed in Sf21 cells

Application

Research Category
Metabolism
Research Sub Category
Cardiovascular Disease

Metabolic Disorders

Biochem/physiol Actions

Protein Target: GCK
Target Sub-Family: STE

Quality

Routinely evaluated by phosphorylation of 0.8 mg/mL myelin basic protein.
Purity was assessed by SDS-PAGE and Coomassie blue staining using 3 μg of active GCK.

Unit Definition

Specific Activity: where one unit of GCK activity is defined as 1nmol phosphate incorporated into 0.8 mg/mL myelin basic protein per minute at 30°C with a final ATP concentration of 100 μM.

Physical form

Purified using glutathione-agarose. Purity 52% by SDS-PAGE and Coomassie blue staining.

Storage and Stability

Stable for 6 months at -70 °C from date of shipment. For maximum recovery of product, centrifuge original vial prior to removing the cap.
Rapidly thaw the vial under cold water and immediately place on ice. Aliquot unused material into pre-chilled microcentrifuge tubes and immediately snap-freeze the vials in liquid nitrogen prior to re-storage at -70 °C.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service