Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

736260

Sigma-Aldrich

1-Aminocyclopropane-2,2,3,3-d4-carboxylic acid

98 atom % D, 98% (CP)

Synonym(s):

ACPC-d4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3D4NO2
CAS Number:
Molecular Weight:
105.13
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

98 atom % D

Assay

98% (CP)

form

powder

mp

229-231 °C (lit.)

mass shift

M+4

SMILES string

[2H]C1([2H])C([2H])([2H])C1(N)C(O)=O

InChI

1S/C4H7NO2/c5-4(1-2-4)3(6)7/h1-2,5H2,(H,6,7)/i1D2,2D2

InChI key

PAJPWUMXBYXFCZ-LNLMKGTHSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

736260-VAR:
736260-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service