338346
Fmoc-Pro-OH
97%
Synonym(s):
N-(9-Fluorenylmethoxycarbonyl)-L-proline, Fmoc-L-proline
About This Item
Recommended Products
Assay
97%
optical activity
[α]20/D −32°, c = 1 in DMF
impurities
≤3% ethyl acetate
mp
117-118 °C (lit.)
SMILES string
OC(=O)[C@@H]1CCCN1C(=O)OCC2c3ccccc3-c4ccccc24
InChI
1S/C20H19NO4/c22-19(23)18-10-5-11-21(18)20(24)25-12-17-15-8-3-1-6-13(15)14-7-2-4-9-16(14)17/h1-4,6-9,17-18H,5,10-12H2,(H,22,23)/t18-/m0/s1
InChI key
ZPGDWQNBZYOZTI-SFHVURJKSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
338346-BULK:
338346-25G:
338346-5G:
338346-PM:
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service