Skip to Content
Merck
All Photos(1)

Documents

Safety Information

577901

Sigma-Aldrich

Merrifield′s peptide resin

50-100 mesh, extent of labeling: 2.5-4.0 mmol/g Cl- loading, 1 % cross-linked with divinylbenzene

Synonym(s):

Chloromethylated polystyrene, Chloromethylated stryene/divinylbenzene copolymer, Chloromethylated styrene-divinylbenzene resin, Copolymer of styrene and divinylbenzene, chloromethylated, Poly(styrene-co-vinylbenzyl chloride-co-divinylbenzene), Polystyrene crosslinked with divinylbenzene, chloromethylated

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

crosslinking

1 % cross-linked with divinylbenzene

reaction suitability

reaction type: Boc solid-phase peptide synthesis

extent of labeling

2.5-4.0 mmol/g Cl- loading

particle size

50-100 mesh

application(s)

peptide synthesis

SMILES string

ClCc1ccccc1.C=Cc2ccccc2.C=Cc3ccc(C=C)cc3

InChI

1S/CH3Cl/c1-2/h1H3

InChI key

NEHMKBQYUWJMIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

577901-1KG:
577901-BULK:
577901-25G:
577901-100G:
577901-VAR:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service