239666
2-(Methylamino)ethanol
≥99%
Synonym(s):
N-Methylethanolamine
About This Item
Recommended Products
vapor density
2.6 (vs air)
vapor pressure
0.5 mmHg ( 20 °C)
Assay
≥99%
autoignition temp.
662 °F
expl. lim.
19.8 %
refractive index
n20/D 1.439 (lit.)
bp
159 °C (lit.)
density
0.935 g/mL at 25 °C (lit.)
SMILES string
CNCCO
InChI
1S/C3H9NO/c1-4-2-3-5/h4-5H,2-3H2,1H3
InChI key
OPKOKAMJFNKNAS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2 - Skin Corr. 1B - STOT RE 2 Oral - STOT SE 3
Target Organs
Kidney,Testes,Liver,spleen,ovary, Respiratory system
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
168.8 °F - closed cup
Flash Point(C)
76 °C - closed cup
Personal Protective Equipment
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
FSL
Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water soluble liquid
ISHL Indicated Name
Substances Subject to be Indicated Names
ISHL Notified Names
Substances Subject to be Notified Names
JAN Code
239666-50ML:
239666-BULK:
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service