Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

223719

Sigma-Aldrich

Sodium hexafluoroarsenate(V)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaAsF6
CAS Number:
Molecular Weight:
211.90
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

Quality Level

Assay

86.9-91.3% (% AsF6 With Tetraphenylarsonium Chloride, gravimetric)

form

powder

reaction suitability

reagent type: catalyst
core: arsenic

SMILES string

[Na+].F[As-](F)(F)(F)(F)F

InChI

1S/AsF6.Na/c2-1(3,4,5,6)7;/q-1;+1

InChI key

NFXMAZFYHDSPPB-UHFFFAOYSA-N

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PDSCL

Poisonous substance

PRTR

Specified Class I Designated Chemical Substances

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

223719-VAR:
223719-BULK:
223719-50G:
223719-10G:4548173118451
223719-10G-PW:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service