Skip to Content
Merck
All Photos(3)

Documents

MABT533

Sigma-Aldrich

Anti-PECAM-1 Antibody, clone 4A8.1

clone 4A8.1, from mouse

Synonym(s):

Platelet endothelial cell adhesion molecule, CD31, EndoCAM, GPIIA′, PECA1, PECAM-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4A8.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... PECAM1(5175)

General description

Platelet endothelial cell adhesion molecule (UniProt P16284; also known as CD31, EndoCAM, GPIIA′, PECA1, PECAM-1) is encoded by the PECAM1 gene (Gene ID 5175) in human. PECAM-1 (CD31) is a 130 kDa cell adhesion molecule that is expressed at the junctions of all continuous endothelia, as well as most bone marrow-derived hematopoietic cells, including platelets, monocytes, granulocytes and some lymphocytes. The human PECAM1 and murine Pecam1 genes both produce multiple spliced isoforms with varying lengths in cytoplasmic domain, with the isoform Long being the major form expressed in human tissues and endothelial cells (ECs), and isoform delta 14-15 being the predominantly expressed murine form. PECAM-1 plays an import role in maintaining vascular integrity and function by mediating ECs interactions with pericytes and smooth muscle cells (SMCs). PECAM-1 is initially produced with a signal peptide (human a.a. 1-27, murine a.a. 1-17) sequence, the removal of which yields the mature protein with a large extracellular region (human a.a. 28-601, mouse a.a. 18-590) composed of six C2-type Ig-like domains, followed by a transmembrane seqment (human a.a. 602-620, mouse a.a. 591-609) and a cytoplasmic tail (human a.a. 621-738, mouse a.a. 610-727) that plays an essential role in PECAM-1-mediated cellular signaling.

Specificity

Clone 4A8.1 was raised against the cytoplasmic domain of PECAM-1 (CD31) isoform Long. Reactivity toward spliced isoforms Delta12, Delta13, Delta14, Delta14-15, and Delta15 with various cytomplasmic domain deletions is possible, but has not not been confirmed.

Immunogen

Epitope: cytoplasmic domain
GST-tagged recombinant human PECAM-1 cytoplasmic domain.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected PECAM-1 in human cerebellum, lung, and pancreas tissue sections.

Western Blotting Analysis: 0.5 µg/mL from a representative lot detected PECAM-1 in 10 µg of Jurkat cell lysate.
Research Category
Cell Structure
This mouse monoclonal Anti-PECAM-1 Antibody, clone 4A8.1, Cat. No. MABT533, is validated for use in Immunohistochemistry (Paraffin) and Western Blotting for the detection of PECAM-1 (CD31).

Quality

Evaluated by Western Blotting in THP-1 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected PECAM-1 in 10 µg of THP-1 human acute monocytic leukemia cell lysate.

Target description

~130 kDa observed. Target band size appears larger than the calculated molecular weights of 79.58/77.52/77.25/77.46/76.30/78.42 kDa (mature isoform Long/Delta12/Delta13/Delta14/Delta14-15/Delta15) and 82.54/80.48/80.21/80.42/79.26/81.38 kDa (isoform Long/Delta12/Delta13/Delta14/Delta14-15/Delta15 pro-form) due to glycosylation. Uncharacterized bands may be observed in some lysate(s).

Linkage

Replaces: 04-1074

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service