HPLC Analysis of Protein Standards (1) on BIOshell™ A400 Protein C4
Conditions | |
---|---|
column | BIOshell A400 Protein C4, 10 cm X 2.1 mm, 3.4 µm particles (66825-U) |
mobile phase | (A) 75:25, [0.1% TFA in water]:[0.1% TFA in acetonitrile] ; (B) 25:75, [0.1% TFA in water]:[0.1% TFA in acetonitrile] |
gradient | 25 to 65% B in 8 min |
flow rate | 0.3 mL/min |
0.3 mL/min | 1430 psi (99 bar) |
sample | protein mixture, g/L ea: lysozyme, 0.1; haptoglobulin, 0.3; conalbumin, 0.3; trypsin inhibitor, 0.3; glutamate dehydrogenase, 0.8; beta-galactosidase, 0.7; ovalbumin, 0.8; lactate dehydrogenase, 0.4 |
injection | 1 µL |
detector | UV, 215 nm |
Description | |
---|---|
Analysis Note | This application demonstrates the utility of the BIOshell A400 Protein C4 column for the reversed-phase chromatography of proteins from a variety of sources and of different molecular weights. |
Categories | Analytical Chromatography, Proteins |
Featured Industry | Life Science and Biopharma |
suitability | application for HPLC |
Materials
Si è verificato un errore inatteso.
Network error: Failed to fetch