Discovery Cyano HPLC Columns
Unique retention and selectivity enables better, faster separations
- Excellent peak shape and efficiency
- Unique selectivity compared to a C18
- Significantly less retentive than C18, reducing the amount of organic needed in the mobile phase
Bonded Phase: | Cyanopropylsilane, endcapped |
Silica: | Spherical, high purity (Fe <20; NA <7; Ca <7; Ti <1; AI <1; Mg <1ppm) |
Particle Size: | 5 µM |
Pore Size: | 180(Å) |
Surface Area: | 200m2/g |
%C: | 4.5% |
Coverage: | 3-4µmoles/m2 |
Ordering Information
5 µM Discovery Cyano HPLC Columns
ID(mm) | Length(cm) | Cat. No. | ||
---|---|---|---|---|
NEW! 2.1 | 2 |
| ||
NEW! 2.1 | 3 |
| ||
2.1 | 5 | 59355-U21 | ||
2.1 | 10 | 569521-U | ||
2.1 | 12.5 | 569524-U | ||
2.1 | 15 | 59356-U21 | ||
NEW! 3.0 | 2 |
| ||
NEW! 3.0 | 3 |
| ||
3.0 | 5 | 59355-U30 | ||
3.0 | 10 | 569522-U | ||
3.0 | 12.5 | 569525-U | ||
3.0 | 15 | 59356-U30 | ||
3.0 | 25 | 59357-U30 | ||
4.0 | 5 | 59355-U40 | ||
4.0 | 10 | 569523-U | ||
4.0 | 12.5 | 569526-U | ||
4.0 | 15 | 59356-U40 | ||
4.0 | 25 | 59357-U40 | ||
NEW! 4.6 | 2 |
| ||
NEW! 4.6 | 3 |
| ||
4.6 | 5 | 59355-U | ||
4.6 | 10 | 569520-U | ||
4.6 | 12.5 | 569527-U | ||
4.6 | 15 | 59356-U | ||
4.6 | 25 | 59357-U |
5 µM Discovery Cyano Validation Packs*
ID(mm) | Length(cm) | Cat. No. |
---|---|---|
2.1 | 5 | 55715-U21 |
2.1 | 10 | 569806-U |
2.1 | 15 | 55717-U21 |
4.6 | 5 | 55715-U |
4.6 | 10 | 569807-U |
4.6 | 15 | 55717-U |
4.6 | 25 | 55719-U |
* Packs include 3 columns, each from a different lof of bonded phase. |
2cm Supelguard Cartridges with Discovery Cyano Packings
How to Contact Supelco
In USA:
Phone: 800-247-6628 or 814-359-3441
fax: 800-325-5052
email: [email protected]
Outside the USA: click here