- Flavonoid constituents from Glycyrrhiza glabra hairy root cultures.
Flavonoid constituents from Glycyrrhiza glabra hairy root cultures.
Phytochemistry (2001-01-05)
W Li, Y Asada, T Yoshikawa
PMID11140606
ABSTRACT
An unusual biflavonoid named licoagrodin was isolated from the hairy root cultures of Glycyrrhiza glabra (Leguminosae) along with three prenylated retrochalcones, licoagrochalcones B, C, D, a prenylated aurone, licoagroaurone and four known prenylated flavonoids, licochalcone C, kanzonol Y, glyinflanin B and glycyrdione A. From the glycosidic fraction, a isoflavone glycoside, licoagroside A, and a maltol glycoside, licoagroside B were isolated together with four known isoflavone glycosides, two flavone C-glycosides, and three other glycosides. Their structures were elucidated on the basis of spectroscopic evidence.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto