Merck
Reductive metabolism of 1,1,1,2-tetrachloroethane and related chloroethanes by rat liver microsomes.
Chemico-biological interactions (1984-10-01)
J A Thompson, B Ho, S L Mastovich
ABSTRACT
The susceptibility of polychlorinated ethanes to reductive metabolism was evaluated by measuring the amount of each compound consumed during anaerobic incubations with rat live microsomes; 1,1,1,2-tetrachloroethane, pentachloroethane and hexachloroethane were metabolized extensively, 1,1,1,2-tetrachloroethane and the trichloroethanes were metabolized very slowly and the dichloroethanes were not metabolized at a detectable rate. The electron affinity of the chloroethanes was determined by measuring electrochemical half-wave reduction potentials. Chloroethanes with an E1/2 of - 1.35 V or less negative were reduced readily in microsomes while those with an E1/2 equal to or more negative than -1.90 V were not good substrates for enzymatic reduction. Metabolites produced from 1,1,1,2-tetrachloroethane in vitro were 1,1-dichloroethylene (DCE) and 1,1,2-trichloroethane (TCEA) and the ratio DCE/TCEA was about 25:1. These conversions were NADPH-dependent and were inhibited by air, CO and metyrapone. In the presence of SKF 525-A, DCE formation was inhibited by 47%. Microsomes from untreated or beta-naphthoflavone-treated rats were 70-90% less active than microsomes from phenobarbital-treated rats. The Km was 0.50 mM and the Vmax was 66 nmol min-1 mg-1 protein for DCE formation. The results are consistent with the proposal that 1,1,1,2-tetrachloroethane is reduced by hepatic cytochrome(s) P-450 to a free radical intermediate which, for the most part, remains closely associated with the enzyme, is reduced further and undergoes beta-elimination of a chloride ion to form DCE. The occurrence of this reductive pathway in vivo was demonstrated by the quantitation of DCE and TCEA in blood from rats treated with 1,1,1,2-tetrachloroethane.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
1,1,1,2-Tetrachloroethane, ReagentPlus®, 99%
Prezzi e disponibilità al momento non sono disponibili