Merck
  • Discovery of orally active carboxylic acid derivatives of 2-phenyl-5-trifluoromethyloxazole-4-carboxamide as potent diacylglycerol acyltransferase-1 inhibitors for the potential treatment of obesity and diabetes.
Discovery of orally active carboxylic acid derivatives of 2-phenyl-5-trifluoromethyloxazole-4-carboxamide as potent diacylglycerol acyltransferase-1 inhibitors for the potential treatment of obesity and diabetes.
Journal of medicinal chemistry (2011-03-19)
Yimin Qian, Stanley J Wertheimer, Mushtaq Ahmad, Adrian Wai-Hing Cheung, Fariborz Firooznia, Matthew M Hamilton, Stuart Hayden, Shiming Li, Nicholas Marcopulos, Lee McDermott, Jenny Tan, Weiya Yun, Liang Guo, Anjula Pamidimukkala, Yingsi Chen, Kuo-Sen Huang, Gwendolyn B Ramsey, Toni Whittard, Karin Conde-Knape, Rebecca Taub, Cristina M Rondinone, Jefferson Tilley, David Bolin
ABSTRACT
Diacylglycerol acyltransferase-1 (DGAT-1) is the enzyme that catalyzes the final and committed step of triglyceride formation, namely, the acylation of diacylglycerol with acyl coenzyme A. DGAT-1 deficient mice demonstrate resistance to weight gain on high fat diet, improved insulin sensitivity, and reduced liver triglyceride content. Inhibition of DGAT-1 thus represents a potential novel approach for the treatment of obesity, dyslipidemia, and metabolic syndrome. In this communication, we report the identification of the lead structure 6 and our lead optimization efforts culminating in the discovery of potent, selective, and orally efficacious carboxylic acid derivatives of 2-phenyl-5-trifluoromethyloxazole-4-carboxamides. In particular, compound 29 (DGAT-1 enzyme assay, IC(50) = 57 nM; CHO-K1 cell triglyceride formation assay, EC(50) = 0.5 μM) demonstrated dose dependent inhibition of weight gain in diet induced obese (DIO) rats (0.3, 1, and 3 mg/kg, p.o., qd) during a 21-day efficacy study. Furthermore, compound 29 demonstrated improved glucose tolerance determined by an oral glucose tolerance test (OGTT).
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
N-Boc-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, 95%