Merck
  • Development of (4-Cyanophenyl)glycine Derivatives as Reversible Inhibitors of Lysine Specific Demethylase 1.
Development of (4-Cyanophenyl)glycine Derivatives as Reversible Inhibitors of Lysine Specific Demethylase 1.
Journal of medicinal chemistry (2017-09-12)
Daniel P Mould, Cristina Alli, Ulf Bremberg, Sharon Cartic, Allan M Jordan, Matthis Geitmann, Alba Maiques-Diaz, Alison E McGonagle, Tim C P Somervaille, Gary J Spencer, Fabrice Turlais, Donald Ogilvie
ABSTRACT
Inhibition of lysine specific demethylase 1 (LSD1) has been shown to induce the differentiation of leukemia stem cells in acute myeloid leukemia (AML). Irreversible inhibitors developed from the nonspecific inhibitor tranylcypromine have entered clinical trials; however, the development of effective reversible inhibitors has proved more challenging. Herein, we describe our efforts to identify reversible inhibitors of LSD1 from a high throughput screen and subsequent in silico modeling approaches. From a single hit (12) validated by biochemical and biophysical assays, we describe our efforts to develop acyclic scaffold-hops from GSK-690 (1). A further scaffold modification to a (4-cyanophenyl)glycinamide (e.g., 29a) led to the development of compound 32, with a Kd value of 32 nM and an EC50 value of 0.67 μM in a surrogate cellular biomarker assay. Moreover, this derivative does not display the same level of hERG liability as observed with 1 and represents a promising lead for further development.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
GSK690, ≥98% (HPLC)