Documenti
V900433
Sigma-Aldrich
N-metil-D-glucamina
Vetec, reagent grade, 99%
Sinonimo/i:
1-desossi-1-(metilamino)-D-glucitolo, Meglumina
About This Item
Formula empirica (notazione di Hill):
C7H17NO5
Numero CAS:
Peso molecolare:
195.21
Beilstein:
385906
Numero CE:
Numero MDL:
Codice UNSPSC:
12352104
ID PubChem:
Grado
reagent grade
Nome Commerciale
Vetec
Saggio
99%
Forma fisica
powder
Temp. autoaccensione
~662 °F
Punto di fusione
129-131.5 °C (lit.)
Stringa SMILE
CNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO
InChI
1S/C7H17NO5/c1-8-2-4(10)6(12)7(13)5(11)3-9/h4-13H,2-3H2,1H3/t4-,5+,6+,7+/m0/s1
MBBZMMPHUWSWHV-BDVNFPICSA-N
Altre note
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Note legali
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Codice della classe di stoccaggio
11 - Combustible Solids
WGK 1
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 2
1 of 2
Thiourea ReagentPlus®, ≥99.0%
Sigma-Aldrich
T7875
Thiourea
Raphaëlle Renard-Penna et al.
Radiology, 275(2), 458-468 (2015-01-20)
To compare the diagnostic performance of the magnetic resonance (MR) imaging-based Prostate Imaging Reporting and Data System (PI-RADS) and a Likert scale in the detection of prostate cancer in a cohort of patients undergoing initial prostate biopsy. This institutional review
Charlotte Manisty et al.
Radiology, 276(3), 732-740 (2015-04-30)
To investigate the pharmacology and potential clinical utility of splenic switch-off to identify understress in adenosine perfusion cardiac magnetic resonance (MR) imaging. Splenic switch-off was assessed in perfusion cardiac MR examinations from 100 patients (mean age, 62 years [age range
Philipp Kickingereder et al.
Neuro-oncology, 17(8), 1139-1147 (2015-03-11)
To analyze the relevance of dynamic susceptibility-weighted contrast-enhanced MRI (DSC-MRI) derived relative cerebral blood volume (rCBV) analysis for predicting response to bevacizumab (BEV) in patients with recurrent glioblastoma (rGB). A total of 127 patients diagnosed with rGB receiving either bevacizumab
Ingrid Millet et al.
Radiology, 270(1), 57-66 (2013-09-14)
To retrospectively compare the kinetic magnetic resonance (MR) imaging characteristics of invasive breast carcinomas with both prognostic tumoral and patient-related parameters. This HIPAA-compliant retrospective study was approved by the institutional review board, and informed consent was waived. From January 2008
Myriam Edjlali et al.
Radiology, 270(1), 261-268 (2013-09-14)
To assess the feasibility of a selective flow-tracking cartographic procedure applied to four-dimensional (4D) flow imaging and to demonstrate its usefulness in the characterization of dural arteriovenous fistulas (DAVFs). Institutional review board approval was obtained, and all patients provided written