Documenti
SU860027
Tappi con ghiera con setti in PTFE/silicone
pkg of 1000 ea
About This Item
Codice UNSPSC:
24122002
Materiali
silver aluminum (with 4 mm center hole)
white red PTFE/silicone
Durezza
45 (Durometer: shore A)
Confezionamento
pkg of 1000 ea
diam. × spessore
8 mm × 1.3 mm
Choose from one of the most recent versions:
Certificati d'analisi (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Servizio Clienti