Documenti
56734-U
Supelco
Stainless Steel 1/16 in. Capillary Tubing
L × O.D. × I.D. 30 cm × 1/16 in. × 0.030 in.
Sinonimo/i:
0.030IN ID SS TUBING, 1/16IN OD, 30CM
About This Item
Codice UNSPSC:
41115700
eCl@ss:
32119290
NACRES:
SB.52
Materiali
white band
Lungh. × D.E. × D.I.
30 cm × 1/16 in. × 0.030 in.
Descrizione generale
316 stainless steel, 1/16 in. O.D. 1 m length is coiled, all other dimensions are straight.
Prodotti consigliati
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificati d'analisi (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Servizio Clienti