Documenti
20392
In-Line Pressure Gauge Kit
About This Item
Codice UNSPSC:
41112500
Parametri
0-100 psi pressure range
tecniche
gas chromatography (GC): suitable
Descrizione generale
Install a pressure gauge in the gas delivery system to measure pressure at any given point. For example, install one upstream and downstream of the High Capacity Gas Purifier. When the pressure drop across the purifier exceeds 10 psi, the converter tube should be replaced. Kit contains:
  • 2 in. (5 cm) diameter steel and copper alloy gauge (0-100 psi)
  • NPT to Swagelok adapter
  • 1/8 in. tee
  • 18 in. (1/2 m) of 1/8 in. copper line
  • assembly and installation instructions
Nuts and ferrules included.
Applicazioni
In-line pressure gauge kit may be used to monitor the column inlet pressure of high-performance liquid chromatography (HPLC) equipment, which is further modified for carbon-dioxide supercritical fluid chromatography (SFC).
Choose from one of the most recent versions:
Certificati d'analisi (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Servizio Clienti
Possiedi già questo prodotto?
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 1
1 of 1
Cleaned Copper Tubing tubing L × O.D. × I.D. 50 ft × 1/8 in. (3.18 mm) × 0.065 in. (1.65 mm)
Supelco
20488
Cleaned Copper Tubing
Modification of conventional high-performance liquid chromatography equipment for use in carbon dioxide supercritical fluid chromatography.
Simpson CR, et al.
Journal of Chromatography A, 371, 109-119 (1986)