Documenti fondamentali
Z741757
4-liter Culture media bottle
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.22
Materiali
polycarbonate
Caratteristiche
Comes with cap, tubing and fixtures.
Produttore/marchio commerciale
IBI ACCFL0008
Categorie correlate
Descrizione generale
The 4L culture media bottle is set up to provide source culture media for an IBI Flow Cell system. The bottle is made of polycarbonate and can be autoclaved. It comes complete with cap, containing fixtures and tubing to connect to the 3-channel or Convertible flow cell systems. - See more at: http://www.ibisci.com/product/accfl0008#sthash.hO4T0B0H.dpuf
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)