Z733768
IKA® ULTRA-TURRAX® disperser tools, plastic
for T-18 basic disperser, diam. 14 mm
About This Item
Materiali
polycarbonate
Caratteristiche
autoclavable
polycarbonate rotor-stator
Confezionamento
pack of 5 ea
Produttore/marchio commerciale
IKA 3451900
Diametro
14 mm
Note legali
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
ULTRA-TURRAX is a registered trademark of IKA-Werke GmbH & Co. KG
Sostituito da