Vai a
V5382
Nalgene® specimen vial
capacity 15 mL
Sinonimo/i:
sample container, sample vial, specimen container, Nalgene 5005-0015, laboratory container, vessel
Autenticati per visualizzare i prezzi organizzativi e contrattuali.
Scegli un formato
Cambia visualizzazione
A voi/SKUDisponibilitàPrezzo
1 pkg
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
219,00 €
Informazioni su questo articolo
NACRES:
NB.22
UNSPSC Code:
41121800
219,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Servizio Tecnico
Hai bisogno di aiuto? Il nostro team di scienziati qualificati è a tua disposizione.
Permettici di aiutarti
material
high-density polyethylene cap, polypropylene copolymer bottle, round translucent bottle
joint
joint threaded neck
sterility
sterile
feature
with cap
packaging
pack of 75 ea
manufacturer/tradename
Nalgene® 5005-0015
capacity
15 mL
General description
Vials of polypropylene, linerless screw closures of high-density polyethylene. For storage of solid specimens. Use in mechanical freezers only. Wide-mouth, shoulderless vial allows easy access to sample. Flat bottom permits self-standing on flat surface. Certified sterile, non-cytotoxic and non-pyrogenic.
Packaging
75 vials per package
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Confronta articoli simili
Visualizza il confronto per intero
Mostra le differenze
1 of 1
Questo articolo
V5257Z359033V4757
Nalgene® specimen vial capacity 15 mL
V5382
Nalgene® specimen vial
material
high-density polyethylene cap, round translucent bottle, polypropylene copolymer bottle
material
colorless tube, conical bottom, high-density polyethylene closure, polypropylene , polypropylene tube
material
colorless tube, conical bottom, polypropylene , polypropylene caps, polypropylene tube, silicone gasket
material
colorless tube, conical bottom, high-density polyethylene closure, polypropylene , polypropylene tube
manufacturer/tradename
Nalgene® 5005-0015
manufacturer/tradename
Nalgene® 5000-0050
manufacturer/tradename
Nalgene® 5000-1020
manufacturer/tradename
Nalgene® 5000-0012
feature
with cap
feature
External thread
feature
External thread
feature
External threads
capacity
15 mL
capacity
5 mL
capacity
1.5 mL
capacity
1.2 mL
joint
joint threaded neck
joint
-
joint
-
joint
-
packaging
pack of 75 ea
packaging
pack of 250 ea (25 x 10 ea)
packaging
pack of 500 ea (20 x 25 ea)
packaging
package of 25 vials x 20, pack of 500 ea (20 x 25ea)
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)