T4793
Nalgene® centrifuge tubes, Round Style 3117
nominal capacity 100 mL
About This Item
Codice UNSPSC:
41121703
Materiali
lipped
polycarbonate
Sterilità
non-sterile
Caratteristiche
RCF 50,000 × g
Confezionamento
pack of 10
Produttore/marchio commerciale
Nalgene®
Capacità nominale
100 mL
Descrizione generale
These high-speed polycarbonate tubes withstand temperatures from -135° to +135°C (-211° to +275°F)
- Designed with round bottoms to fit high-speed rotors
- Molding technique produces uniform wall thickness for increased strength
- Uniform diameter (no taper) for better rotor fit and force tolerance
- Lip allows easier retrieval from rotor cavities
- Economical choice - may also be used as test tubes
- Longer-lasting than polycarbonate tubes under conditions of repeated autoclaving
- Autoclavable/Transparent
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)