Documenti fondamentali
T4414
Sigma-Aldrich
Silicone rubber laboratory tubing
I.D. × O.D. 3/8 in. × 5/8 in.
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.35
Diam. int. × diam. est.
3/8 in. × 5/8 in.
Misura parete
1/8 in.
Descrizione generale
Translucent and flexible. Temp. range: -60 to 460°F (-51 to 238°C). Do not use with concentrated acids or dilute sodium hydroxide. Suitable for high-temperature applications.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)