Key Documents
RFA452411-EP
Rotor for Eppendorf® 5430/5430R Centrifuges
F-45-24-11, non aerosol tight, to hold, 24 x 1.5/2.0 mL tubes, lid: no
About This Item
Codice UNSPSC:
41103909
Caratteristiche
lid: no
to hold 24 x 1.5/2.0 mL tubes
fixed angle rotor
non aerosol tight
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Eppendorf® 022639471 (NA)
Eppendorf® 5702746002 (EU)
Parametri
1,770 × g max. RCF
tecniche
centrifugation: suitable
Compatibilità
for use with centrifuge models 5430/5430R
Categorie correlate
Note legali
Eppendorf is a registered trademark of Eppendorf AG