Vai a
MTCB1212
MTC™ Bio MultiBox™ for Western Blotting
2 compartments, L × W × D 85 mm (311/32 in.) × 85 mm (311/32 in.) × 30 mm (13/16 in.), for Novex Mini (8x8cm), Bio-Rad Mini (7x8cm) and others
Sinonimo/i:
gel processing box, multi gel processing box, MTC Bio B1212, western blot apparatus, western blot chamber
Autenticati per visualizzare i prezzi organizzativi e contrattuali.
Scegli un formato
Cambia visualizzazione
Voi/SKUDisponibilitàPrezzo
Informazioni su questo articolo
NACRES:
NB.15
UNSPSC Code:
41121800
Manufacturer/tradename:
MTC™ Bio B1212
Material:
polystyrene
Packaging:
pack of 6 ea
Prezzi e disponibilità al momento non sono disponibili
Servizio Tecnico
Hai bisogno di aiuto? Il nostro team di scienziati qualificati è a tua disposizione.
Permettici di aiutarti
material
polystyrene
packaging
pack of 6 ea
manufacturer/tradename
MTC™ Bio B1212
L × W × D
85 mm (311/32 in.) × 85 mm (311/32 in.) × 30 mm (13/16 in.)
General description
MultiBox™, 2 compartments, 85 x 85 x 30mm each (311/32 x 311/32 x 13/16 in.), for Novex Mini (8x8cm), Bio-Rad Mini (7x8cm) and others, 6/pk
The MultiBox™ Western Blotting Box provides the ease and convenience of using one box to process multiple gels, allowing users to compare results side by side in real time.
Features and Benefits
  • A perfect box for every blot
  • Reduce antibody usage
  • Hinged lid minimizes evaporation
  • For halves, wholes, strips or minis
Legal Information
MTC is a trademark of MTC Bio
MultiBox is a trademark of MTC Bio
Confronta articoli simili
Visualizza il confronto per intero
Mostra le differenze
1 of 1
Questo articolo
MTCB1204MTCB1203MTCB1206
description
2 compartments, L × W × D 85 mm (311/32 in.) × 85 mm (311/32 in.) × 30 mm (13/16 in.), for Novex Mini (8x8cm), Bio-Rad Mini (7x8cm) and others
description
4 compartments, L × W × D 38 mm (1 1/2 in.) × 71 mm (2 13/16 in.) × 28 mm (1 1/8 in.), for various gels including half-mini protein gels
description
4 compartments, L × W × D 32 mm (11/4 in.) × 52 mm (2 5/16 in.) × 28 mm (1 1/8 in.), for various gels
description
6 compartments, L × W × D 33 mm (4 in.) × 103 mm (1 1/4 in.) × 35 mm (1 3/8 in.)
manufacturer/tradename
MTC™ Bio B1212
manufacturer/tradename
MTC™ Bio B1204
manufacturer/tradename
MTC™ Bio B1203
manufacturer/tradename
MTC™ Bio B1206
material
polystyrene
material
polystyrene
material
polystyrene
material
polystyrene
packaging
pack of 6 ea
packaging
pack of 6 ea
packaging
pack of 6 ea
packaging
pack of 4 ea
L × W × D
85 mm (311/32 in.) × 85 mm (311/32 in.) × 30 mm (13/16 in.)
L × W × D
38 mm (1 1/2 in.) × 71 mm (2 13/16 in.) × 28 mm (1 1/8 in.)
L × W × D
32 mm (11/4 in.) × 52 mm (2 5/16 in.) × 28 mm (1 1/8 in.)
L × W × D
33 mm (4 in.) × 103 mm (1 1/4 in.) × 35 mm (1 3/8 in.)
Still not finding the right product?
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)