Scegli un formato
| A voi/SKU | Disponibilità | Prezzo |
|---|---|---|
1 ea | Disponibile per la spedizione OGGIdaSchnelldorf Distribution | 181,00 € |
Informazioni su questo articolo
181,00 €
Disponibile per la spedizione OGGIDettagli
material
clear acrylic
feature
to hold 8 x 80 well racks
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS23263B
L × W × H
310 mm × 236 mm × 122 mm
General description
Features and Benefits
- Holds up to eight 80-well microtube racks filled with either 1.5 or 2.0 mL microtubes
- Plenty of room to slide the racks in and out
1 of 1
Questo articolo | |||
|---|---|---|---|
| description to hold, 8 x 80 well racks, clear acrylic | description to hold, 8 x 80 well racks, clear acrylic rack | description Holds 15 x slide boxes, clear acrylic | description - |
| feature to hold 8 x 80 well racks | feature to hold 8 x 80 well racks | feature Holds 15 x slide boxes | feature Holds 6 x hybridization bottles |
| manufacturer/tradename Heathrow Scientific HS23263B | manufacturer/tradename Heathrow Scientific HS23263A | manufacturer/tradename Heathrow Scientific HS159890B | manufacturer/tradename WHEATON® 805015 |
| material clear acrylic | material clear acrylic rack | material clear acrylic | material - |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging - | packaging - |
| L × W × H 310 mm × 236 mm × 122 mm | L × W × H 157 mm × 236 mm × 234 mm | L × W × H - | L × W × H - |
Scegli una delle versioni più recenti:
Possiedi già questo prodotto?