Scegli un formato
| Voi/SKU | Disponibilità | Prezzo |
|---|---|---|
1 ea | Per informazioni sulla disponibilità, contatta il Servizio Clienti. | 190,00 € |
Informazioni su questo articolo
190,00 €
material
orange , white
feature
adjustable, single-channel
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS120100
volume measuring range
5-50 μL
General description
Features and Benefits
- Internal metal parts for increased accuracy and calibration stability, and long life
- Ergonomic Slight downward slant to tip ejector button reduces hand fatigue, incidence of repetitive strain injury (RSI)
- Easy volume adjustment with locking tab to prevent accidental change
- Separate tip ejector
- Large digit volume display
- Color coded and labeled with volume range
- Both top and side have colored coded components for quick visual confirmation
- CE certified and Manufactured in accordance with ISO08655 and GLP regulations
- Autoclavable
Legal Information
1 of 1
Questo articolo | |||
|---|---|---|---|
| description adjustable, volume 5-50 μL, orange, single-channel | description adjustable, volume 0.1-2 μL, green, single-channel | description adjustable, volume 2-20 μL, light purple, single-channel | description adjustable, volume 1000-10000 μL, red, single-channel |
| material orange , white | material white , green | material light purple , white | material white , red |
| volume measuring range 5-50 μL | volume measuring range 0.1-2 μL | volume measuring range 2-20 μL | volume measuring range 1000-10000 μL |
| manufacturer/tradename Heathrow Scientific HS120100 | manufacturer/tradename Heathrow Scientific HS120097 | manufacturer/tradename Heathrow Scientific HS120099 | manufacturer/tradename Heathrow Scientific HS120106 |
| feature adjustable, single-channel | feature single-channel, adjustable | feature adjustable, single-channel | feature single-channel, adjustable |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
Still not finding the right product?
Explore all of our products under Pearl™ Pipette
Scegli una delle versioni più recenti:
Possiedi già questo prodotto?