Documenti
GN782096
Greiner microplate
1536 well, HiBase, flat μClear® bottom, medium binding, black
Sinonimo/i:
1536 well microplate, 1536 well plate, cell culture plate
About This Item
Codice UNSPSC:
41121800
Confezionamento
pack of 60 ea
Produttore/marchio commerciale
Greiner 782096
Disponibilità
available only in China
Descrizione generale
  • Greiner microplates are ideal for storing active agents, patient samples, DNA/RNA or stock cultures.
  • Polypropylene material can withstand a high temperature (-196 to +121°C) and are chemically resistant.
  • Nonpyrogenic plates are free of detectable DNase, RNase, human DNA.
  • Features alphanumeric well coding for identification.
  • Sealable with adhesive films and heat sealers for long-term and secure sealing.
Note legali
μClear is a registered trademark of Greiner Bio-One GmbH