GN655207
Greiner microplate
96 well, flat bottom, chimney style, white
Sinonimo/i:
96 well microplate, 96 well plate, cell culture plate
About This Item
Codice UNSPSC:
41121800
Materiali
white
Confezionamento
pack of 100 ea
Produttore/marchio commerciale
Greiner 655207
Disponibilità
available only in China
Misura
96 wells
Compatibilità
suitable for (cell culture)
Descrizione generale
- Greiner microplates are ideal for storing active agents, patient samples, DNA/RNA or stock cultures.
- Polypropylene material can withstand a high temperature (-196 to +121°C) and are chemically resistant.
- Nonpyrogenic plates are free of detectable DNase, RNase, human DNA.
- Features alphanumeric well coding for identification.
- Sealable with adhesive films and heat sealers for long-term and secure sealing.
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)