Scegli un formato
| Voi/SKU | Disponibilità | Prezzo |
|---|---|---|
4 x 8 ea | Per informazioni sulla disponibilità, contatta il Servizio Clienti. | 747,00 € |
Informazioni su questo articolo
747,00 €
material
screw top blue high-density polyethylene cap, straight neck , straight neck transparent polystyrene flask
sterility
sterile
packaging
case of 32 ea
manufacturer/tradename
Nunc 132913
technique(s)
cell culture: suitable
flask working volume
200 mL
surface area
500 cm2 , 500 cm2 , culture area (cm2) per flask
working volume
200 mL
binding type
Nunclon Delta surface
General description
Features and Benefits
- Three parallel growth surfaces provide a total culture area of 500 cm2
- External dimensions of a 175 cm2 standard flask
- Ideal for scale-up cell culture applications
- Extra caps in every carton
- Nunclon™ Δ surface ensures consistent optimal cell attachment and growth
- Barcoded with large Code 128
Legal Information
1 of 1
Questo articolo | |||
|---|---|---|---|
| description Culture flask, surface area 500 cm2, with cap filtered, straight neck, sterile, case of 32 ea | description Nunclon® Delta, surface area (500 cm2), filter cap, straight neck, sterile, case of 32 ea | description Culture flask, surface area 500 cm2, with solid cap, straight neck, sterile, case of 32 ea | description Nunclon® Δ surface treatment, 75 cm2 angled neck, filter cap, 100/cs |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile; γ-irradiated |
| manufacturer/tradename Nunc 132913 | manufacturer/tradename Nunc 132920 | manufacturer/tradename Nunc 132867 | manufacturer/tradename Nunc 156499 |
| binding type Nunclon Delta surface | binding type Nunclon Delta surface | binding type Nunclon Delta surface | binding type Nunclon Delta surface |
| material screw top blue high-density polyethylene cap, straight neck , straight neck transparent polystyrene flask | material straight neck , straight neck transparent polystyrene flask, screw top blue high-density polyethylene cap | material screw top blue high-density polyethylene cap, straight neck transparent polystyrene flask | material screw top blue high-density polyethylene cap (filter), canted neck transparent polystyrene flask |
| working volume 200 mL | working volume 200 mL | working volume 200 mL | working volume - |
| surface area 500 cm2 , 500 cm2 , culture area (cm2) per flask | surface area (500 cm2) , 500 cm2 , culture area (cm2) per flask | surface area 500 cm2 , 500 cm2 , culture area (cm2) per flask | surface area 75 cm2 |
Still not finding the right product?
Explore all of our products under Nunc® TripleFlasks, Nunclon® Δ Surface
Scegli una delle versioni più recenti:
Possiedi già questo prodotto?