Vai a
F8145
Carestream® electron image film SO-163
Autenticati per visualizzare i prezzi organizzativi e contrattuali.
Informazioni su questo articolo
NACRES:
MA.01
UNSPSC Code:
41105341
sheet size
3 1/4 in. × 4 in.
storage temp.
2-8°C
General description
Fine-grain, blue-sensitive emulsion on 7-mil ESTAR base, notched for emulsion identification. Tear- and abrasion-resistant, dimensionally stable. Greater electron exposure and development latitudes than 4489.
Legal Information
Kodak is a registered trademark of Eastman Kodak Company
Still not finding the right product?
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)