Documenti fondamentali
F5051
Corning® FiltrEX 96 well filter plates
96 well, PVDF membrane, pore size 0.2 μm
About This Item
Codice UNSPSC:
41104900
Materiali
PVDF membrane
polystyrene
rectangular bottom white polystyrene wells
white polystyrene plate
Sterilità
sterile
Caratteristiche
lid: no
plate format: 96 well filter plate, standard
Confezionamento
pack of 10
case of 50
Produttore/marchio commerciale
Corning 3505
Volume massimo
350 μL
Volume d’esercizio
350 μL
Dimensione pori
0.2 μm
Descrizione generale
Glass fiber filter plates (CLS3510, CLS3511) can be used for a variety of applications, including plasmid isolation, DNA purification, or receptor/ligand binding assays. The low-binding hydrophilic PVDF membrane filter plates (CLS3504, CLS3505) are suitable for protein kinase assays or for bead and resin based separation assays.
Note legali
Corning is a registered trademark of Corning, Inc.
FiltrEX is a trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)