EP6731011085
Eppendorf® CellXpert® C170i CO2 Incubator
Oxygen control, inner door with 8 door segments, handle right side, AC/DC input 120 V, US 2-pin plug (US/JP/South America)
About This Item
Codice UNSPSC:
41104405
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Eppendorf® 6731011085
Categorie correlate
Descrizione generale
CellXpert® C170i, inner door with 8 door segments, handle right side, 1 – 20 % oxygen control, 100 – 127 V/50 – 60 Hz (US/JP/South America)
Caratteristiche e vantaggi
- 1 – 20 % oxygen control
- handle right side
- inner door with 8 door segments
- 180° HTD (standard with C170i)
- including water tray and four shelves (stainless steel)
- 100 – 127 V, 50 – 60 Hz
Note legali
CellXpert is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)