Documenti
EP5810761000
Eppendorf® Adapter for 250 mL Rectangular Bucket
for use with 80-120 mL tubes, set of 2 pieces
About This Item
Codice UNSPSC:
41103911
NACRES:
NB.82
Caratteristiche
for use with 80-120 mL tubes
Confezionamento
set of 2 pieces
Produttore/marchio commerciale
Eppendorf® 5810761000
tecniche
centrifugation: suitable
Compatibilità
for use with 250 mL rectangular buckets
Note legali
Eppendorf is a registered trademark of Eppendorf AG