Documenti
EP022653121
Rotor for Eppendorf® 5424/5424R Centrifuges
strip rotor, F-45-64-5-PCR, non aerosol tigh, to hold, 4 x 8 tube PCR strips
About This Item
Codice UNSPSC:
41103909
Caratteristiche
to hold 4 x 8 tube PCR strips
strip rotor, F-45-64-5-PCR, non aerosol tigh
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Eppendorf® 022653121 (North America)
Parametri
15,000 max. RCF (18,615 xg)
tecniche
PCR: suitable
centrifugation: suitable
Compatibilità
for use with centrifuge models 5424/5424R
Note legali
Eppendorf is a registered trademark of Eppendorf AG