Merck
C1437
CryoSleeve clear PVC cane sleeve
L 273 mm
Sinonimo/i:
cryogenic sleeve, cryostorage sleeve, cryovial sleeve, dewar sleeve, liquid nitrogen sleeve, Nalgene 5016-0001
Autenticati per visualizzare i prezzi organizzativi e contrattuali.
Scegli un formato
Cambia visualizzazione
Informazioni su questo articolo
NACRES:
NB.22
UNSPSC Code:
41103029
Servizio Tecnico
Hai bisogno di aiuto? Il nostro team di scienziati qualificati è a tua disposizione.
Permettici di aiutarti
packaging
pack of 100 ea
manufacturer/tradename
Nalgene 5016-0001
L
273 mm
General description
Give your cryogenic storage tubes and vials extra protection, such as when shipping or storing in liquid or vapor phase nitrogen with this storage accessory. The CryoSleeve slide over any Nunc or Nalgene Cryocane and prevent tubes from falling out of the cryocane and into the bottom of the liquid nitrogen dewar but do not prevent infiltration of liquid nitrogen. Cryogenic tubes mounted in the cane must first be sealed inside tubing prior to applying the if liquid phase submersion is required.
Features and Benefits
  • For cryogenic storage tubes and vials providing extra protection.
  • For use when shipping or storing in liquid or vapor phase nitrogen.
Legal Information
CryoCane is a trademark of Thermo Fisher Scientific or its subsidiaries
CryoFlex is a trademark of Thermo Fisher Scientific or its subsidiaries
CryoSleeve is a trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)