Documenti fondamentali
AXYPCR96FSY
PCR microplate, full skirt
size 96 wells, conical bottom yellow polypropylene wells, pkg of 5x10plates/cs
Sinonimo/i:
96 multiwell plate
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.22
Materiali
conical bottom yellow polypropylene wells
Sterilità
non-sterile
Produttore/marchio commerciale
Axygen Inc PCR-96-FS-Y
Corning
Confezionamento
pkg of 5x10plates/cs
Volume massimo
200 μL
Misura
96 wells
Volume d’esercizio
200 μL
Tipo di binding
non-treated surface