Documenti fondamentali
AXYHGB1516MC2
Corning® Axygen® 16 well MC comb for use with 15 cm gel system
thickness (2.0 mm)
Sinonimo/i:
gel system accessory
About This Item
Codice UNSPSC:
41105300
NACRES:
NB.15
Produttore/marchio commerciale
Corning HGB15-16MC-2
Spessore
(2.0 mm)
Note legali
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.