Documenti fondamentali
UBBEL04NC
Ubbelohde Viscometer for Transparent Liquids
Size 1, Nom. Constant 0.01, Range 2 to 10 cSt, (non certified)
About This Item
Codice UNSPSC:
41116107
NACRES:
NA.24
Produttore/marchio commerciale
Paragon Scientific Ltd
applicazioni
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
Descrizione generale
This is the non certified version of this viscometer. If you wish to order a certifed viscometer, please refer to the corresponding product UBBEL04UKC
Applicazioni
Ubbelohde Viscometer may be used for the determination of kinematic viscosity of transparent Newtonian liquids according to ASTM D 445, ISO 3104, IP 71, and BS188.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)