Documenti fondamentali
CFR02NC
Cannon-Fenske Routine Viscometer for Transparent Liquids
Size 50, Nom. Constant 0.004, Range 0.8 to 4 cSt, (non certified)
About This Item
Codice UNSPSC:
41116107
Produttore/marchio commerciale
Paragon Scientific Ltd
applicazioni
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
Descrizione generale
This is the non certified version of this viscometer. If you wish to order a certifed viscometer, please refer to the corresponding product CFR02UKC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)