Vai a
324893
Epigenetic Regulators Panel - Acetylation - Calbiochem
Autenticati per visualizzare i prezzi organizzativi e contrattuali.
Informazioni su questo articolo
NACRES:
NA.32
UNSPSC Code:
51111800
Form:
solid
Storage condition:
OK to freeze
form
solid
Quality Segment
storage condition
OK to freeze
solubility
soluble (DMSO (supplied). Please refer to information provided on the CD for reconstitution of individual inhibitors.)
storage temp.
−20°C
General description
A panel containing 18 potent and selective small molecule regulators that is useful for the study of epigenetic acetylation process. Supplied with a data sheet. Epigenetic Regulators Panel - Methylation (Cat. No. 324894) is also available.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)