Z683892
Eppendorf® Research® plus pipette, 3-pack
Option 2
Sinonimo/i:
Adjustable volume pipette
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.01
Prodotti consigliati
Caratteristiche
autoclavable
electronic: no
single-channel
Confezionamento
pack of 3 ea
Produttore/marchio commerciale
Eppendorf® 3123000918
Volume
1000 μL
Descrizione generale
3 pipettes (2–20 μL, 20–200 μL, 100–1,000 μL) + 3 reusable epT.I.P.S.® boxes with 96 pipette tips each.
Comfortable, high precision pipetting is a result of perfect teamwork between pipette and tip. Research plus with PhysioCare Concept® are available in packs combining Eppendorf Research plus pipettes and epT.I.P.S.® pipette tips at a special price.
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Research is a registered trademark of Eppendorf AG
PhysioCare Concept is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)