Key Documents
Z645826
Chemical spill cushions
size medium
Sinonimo/i:
chemical cleanup, spill cleanup, spill safety
About This Item
Codice UNSPSC:
47131905
NACRES:
NB.35
Misura
medium
Descrizione
each cushion absorbs 4.5 Litres
Produttore/marchio commerciale
Clear-spill CCM
Disponibilità
not available in ROW, USA and Canada
Lungh. × largh.
30 cm × 35 cm
Descrizione generale
Cushions can be used with drip pans or in conjection with pads and socks for absorbing larger spills.