Z301167
Quickfit® expansion adapter
top female joint: ST/NS 34/35, bottom male joint: ST/NS 24/29
Sinonimo/i:
XA53
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
Giunto
female joint: ST/NS top34/35
male joint: ST/NS bottom24/29
Produttore/marchio commerciale
Quickfit XA 53
Disponibilità
not available in USA and Canada
not available in USA and Canada
Descrizione generale
These adapters connect items with larger cone to a piece of glassware with a smaller size socket.
Note legali
Quickfit is a registered trademark of SciLabware Limited
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)