Documenti fondamentali
Z217980
Supelco
Michel-Miller column coupling to end fittings
size 25 mm
About This Item
Codice UNSPSC:
12352200
NACRES:
SB.52
Produttore/marchio commerciale
Ace Glass 584049
tecniche
LPLC: suitable
Misura
25 mm
Descrizione generale
PTFE; for coupling column to end fittings of the same internal diameter. Size refers to inside diameter of threads. Includes two FETFE® O-rings.
Note legali
FETFE is a registered trademark of Ace Glass, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)