Z217972
Michel-Miller column coupling to end fittings
size 15 mm
About This Item
Produttore/marchio commerciale
Ace Glass 584047
tecniche
LPLC: suitable
Misura
15 mm
Descrizione generale
PTFE; for coupling column to end fittings of the same internal diameter. Size refers to inside diameter of threads. Includes two FETFE® O-rings.
Note legali
FETFE is a registered trademark of Ace Glass, Inc.